Få mere at vide
Deacetyl Linezolid, TRC
Artikelnummer. 30351779
Skift visning
Mængde:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30351779 100 mg
30368344 10 mg
30363203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30351779 Leverandør Toronto Research Chemicals Leverandørnr. TRCD198960100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 168828-90-8
Molekylær formel C14 H18 F N3 O3
Molekylvægt (g/mol) 295.31
Synonym 2-Oxazolidinone, 5-(aminomethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-, (5S)-, 2-Oxazolidinone, 5-(aminomethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-, (S)-, (-)-(S)-5-(Aminomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one, (5S)-5-(Aminomethyl)-3-(3-fluoro-4-morpholinophenyl)-1,3-oxazolidin-2-one, (5S)-5-(Aminomethyl)-3-[3-fluoro-4-(morpholin-4-yl)phenyl]-1,3-oxazolidin-2-one, (S)-5-Aminomethyl-3-(3-fluoro-4-morpholinophenyl)-1,3-oxazolidin-2-one, (S)-5-Aminomethyl-3-[3-fluoro-4-(morpholin-4-yl)phenyl]oxazolidin-2-one, (S)-N-[[3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]amine, PNU 105868, Linezolid Related Compound C (USP)
IUPAC navn (5S)-5-(aminomethyl)-3-(3-fluoro-4-morpholin-4-ylphenyl)-1,3-oxazolidin-2-one
SMIL NC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2
Tekniske data
Kemisk navn eller materiale Desacetyllinezolid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C14H18FN3O3/c15-12-7-10(18-9-11(8-16)21-14(18)19)1-2-13(12)17-3-5-20-6-4-17/h1-2,7,11H,3-6,8-9,16H2/t11-/m0/s1
Formel vægt 295.1332
Procent renhed >95%