Få mere at vide
Deacetamidine Cyano Dabigatran-d3 Ethyl Ester, TRC
Artikelnummer. 30359598
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30359598 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359598 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19565210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deacetamidine Cyano Dabigatran-d3 Ethyl Ester
CAS 1346597-93-0
Molekylær formel C27H23D3N6O3
Mængde 10 mg
Synonym N-[[2-[[(4-Cyanophenyl)amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-beta-alanine Ethyl Ester Ethanedioate
InChI formel InChI=1S/C27H26N6O3/c1-3-36-26(34)13-15-33(24-6-4-5-14-29-24)27(35)20-9-12-23-22(16-20)31-25(32(23)2)18-30-21-10-7-19(17-28)8-11-21/h4-12,14,16,30H,3,13,15,18H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])n1c(CNc2ccc(cc2)C#N)nc3cc(ccc13)C(=O)N(CCC(=O)OCC)c4ccccn4
IUPAC navn ethyl 3-[[2-[(4-cyanoanilino)methyl]-1-(trideuteriomethyl)benzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate
Molekylvægt (g/mol) 485.55
Formel vægt 485.2255