Få mere at vide
Deacetamidine Cyano Dabigatran-d3 Ethyl Ester Oxalate, TRC
Artikelnummer. 30363524
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30363524 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363524 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19566250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deacetamidine Cyano Dabigatran-d3 Ethyl Ester Oxalate
CAS 1794780-07-6
Molekylær formel C29H25D3N6O7
Mængde 50 mg
Synonym N-[[2-[[(4-Cyanophenyl)amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-beta-alanine Ethyl Ester Ethanedioate
InChI formel InChI=1S/C27H26N6O3.C2H2O4/c1-3-36-26(34)13-15-33(24-6-4-5-14-29-24)27(35)20-9-12-23-22(16-20)31-25(32(23)2)18-30-21-10-7-19(17-28)8-11-21;3-1(4)2(5)6/h4-12,14,16,30H,3,13,15,18H2,1-2H3;(H,3,4)(H,5,6)/i2D3;
SMIL [2H]C([2H])([2H])n1c(CNc2ccc(cc2)C#N)nc3cc(ccc13)C(=O)N(CCC(=O)OCC)c4ccccn4.OC(=O)C(=O)O
IUPAC navn ethyl 3-[[2-[(4-cyanoanilino)methyl]-1-(trideuteriomethyl)benzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate;oxalic acid
Molekylvægt (g/mol) 575.59
Formel vægt 575.2208