Få mere at vide
Deacetamide Linezolid Azide, TRC
Artikelnummer. 30368484
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368484 10 mg
30358177 50 mg
30351522 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368484 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19560010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 168828-84-0
Molekylær formel C14 H16 F N5 O3
Molekylvægt (g/mol) 321.31
Synonym (R)-5-(Azidomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one, Linezolid Related Compound A, Deacetamide Linezolid Azide, 2-Oxazolidinone, 5-(azidomethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-, (5R)-, 2-Oxazolidinone, 5-(azidomethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-, (R)-, (R)-5-Azidomethyl-3-[3-fluoro-4-(morpholin-4-yl)phenyl]oxazolidin-2-one, (R)-[N-[3-Fluoro-4-(morpholin-4-yl)phenyl]-2-oxo-5-oxazolidinyl]methyl azide
IUPAC navn (5R)-5-(azidomethyl)-3-(3-fluoro-4-morpholin-4-ylphenyl)-1,3-oxazolidin-2-one
SMIL Fc1cc(ccc1N2CCOCC2)N3C[C@H](CN=[N+]=[N-])OC3=O
Tekniske data
Kemisk navn eller materiale (R)-5-(Azidomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one
Mængde 10 mg
InChI formel InChI=1S/C14H16FN5O3/c15-12-7-10(1-2-13(12)19-3-5-22-6-4-19)20-9-11(8-17-18-16)23-14(20)21/h1-2,7,11H,3-6,8-9H2/t11-/m0/s1
Formel vægt 321.1237