Få mere at vide
De(2,3-dihydroxy) Nadolol Hydrochloride, TRC
Artikelnummer. 30361104
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30361104 50 mg
1 options
Artikelnummer. 30361104 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22817050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-1-[(1,1-Dimethylethyl)amino]-3-[(5,6,7,8-tetrahydronaphtalen-1-yl)oxy]propan-2-ol Hydrochloride
CAS 15148-92-2
Molekylær formel C17 H27 N O2 . Cl H
Mængde 50 mg
Synonym 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[(5,6,7,8-tetrahydro-1-naphthalenyl)oxy]-, hydrochloride (1:1), 2-Propanol, 1-(tert-butylamino)-3-[(5,6,7,8-tetrahydro-1-naphthyl)oxy]-, hydrochloride (8CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[(5,6,7,8-tetrahydro-1-naphthalenyl)oxy]-, hydrochloride (9CI), (2RS)-1-[(1,1-Dimethylethyl)amino]-3-[(5,6,7,8-tetrahydronaphtalen-1-yl)oxy]propan-2-ol Hydrochloride, Nadolol Imp. G (EP) as Hydrochloride, 1-[(1,1-Dimethylethyl)amino]-3-[(5,6,7,8-tetrahydro-1-naphthalenyl)oxy]-2-propanol hydrochloride, De(2,3-dihydroxy) Nadolol hydrochloride, De(2,3-dihydroxy)nadolol hydrochloride
InChI formel InChI=1S/C17H27NO2.ClH/c1-17(2,3)18-11-14(19)12-20-16-10-6-8-13-7-4-5-9-15(13)16;/h6,8,10,14,18-19H,4-5,7,9,11-12H2,1-3H3;1H
SMIL Cl.CC(C)(C)NCC(O)COc1cccc2CCCCc12
IUPAC navn 1-(tert-butylamino)-3-(5,6,7,8-tetrahydronaphthalen-1-yloxy)propan-2-ol;hydrochloride
Molekylvægt (g/mol) 313.86
Formel vægt 313.1809