missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ DDX17 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16361535
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16361535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16361535 Leverandør Invitrogen™ Leverandørnr. PA584585

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: GGRSRYRTTS SANNPNLMYQ DECDRRLRGV KDGGRRDSAS YRDRSETDRA GYANGSGYGS PNSAFGAQAG QYTYGQ Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an ATPase activated by a variety of RNA species, but not by dsDNA. This protein, and that encoded by DDX5 gene, are more closely related to each other than to any other member of the DEAD box family. This gene can encode multiple isoforms due to both alternative splicing and the use of alternative translation initiation codons, including a non-AUG (CUG) start codon.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen DDX17
Applikationer ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 0.2 mg/mL
Konjugeret Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX17
Genadgangsnr. Q92841
Gene Alias 2610007K22Rik; A430025E01Rik; AI047725; C80929; DDX17; DEAD (Asp-Glu-Ala-Asp) box helicase 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 46; DEAD box polypeptide 17; DEAD box protein 17; DEAD box protein p72; DEAD box protein p82; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD); DEAD/H box 17; DEAD-box helicase 17; DKFZp761H2016; Gm926; P72; Probable ATP-dependent RNA helicase DDX17; RH70; RNA-dependent helicase p72; RP3-434P1.1
Gensymboler DDX17
Værtsarter Rabbit
Immunogen Recombinant protein corresponding to Human DDX17. Recombinant protein control fragment (Product #RP-103492).
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 10521
Målarter Human
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.