missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DDR2, GST Tag Recombinant Protein
DDR2 is a member of a subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain.
Brand: Invitrogen™ PR7167A
Additional Details : Weight : 0.01000kg
Description
DDR1 and DDR2 have been shown to be potently inhibited by Gleevec.
Specifications
NP_006173, S642A | |
79.8 kDa | |
DDR2, TK, NTRKR3 | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
Oncology, Signal Transduction |
Kinase Assay | |
100μg | |
DDR Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |