Få mere at vide
Tocris Bioscience™ DDR1-IN-1 dihydrochloride
GREENER_CHOICE
Selective DDR1 inhibitor
Brand:  Tocris Bioscience™ 5077/10
Encompass
Produktkode 15766615
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
DDR1-IN-1 dihydrochloride is a selective discoidin domain receptor 1 (DDR1) tyrosine kinase inhibitor (IC50 values are 105 and 413 nM for DDR1 and DDR2 respectively). Inhibits integrin-induced DDR1 autophosphorylation in an osteosarcoma cell line.
Tekniske data
Tekniske data
>98%
1780303-76-5
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only