missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR1 Antibody (3E3), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81073H
Additional Details : Weight : 0.00970kg
Description
DDR1 Monoclonal specifically detects DDR1 in Human samples. It is validated for Western Blot, ELISA.Specifications
DDR1 | |
Monoclonal | |
HRP | |
PBS | |
DDR1 | |
Recombinant human DDR1 extracellular domain. | |
0.1 mL | |
Primary | |
This antibody binds specifically to the extracellular domain of human DDR1. | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, ELISA | |
3000 | |
Western Blot, ELISA | |
CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
Rabbit | |
Protein A purified | |
RUO | |
780 | |
Human | |
Purified |