missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ DDIT4L Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17161834 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17161834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17161834 Leverandør Invitrogen™ Leverandørnr. PA5113976

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: ESTCQNLVKM LENCLSKSKQ TKLGCSKVLV PEKLTQRIAQ DVLRLSSTEP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

DDIT4L inhibits cell growth by regulating the TOR signaling pathway upstream of the TSC1-TSC2 complex and downstream of AKT1.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen DDIT4L
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 2% sucrose and 0.09% sodium azide
Gene DDIT4L
Genadgangsnr. Q96D03
Gene Alias 1700037B15Rik; 1700108M02Rik; Ddit4l; DNA damage inducible transcript 4 like; DNA damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like; DNA-damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like protein-like; HIF-1 responsive protein RTP801-like; homolog of mouse SMHS1; protein regulated in development and DNA damage response 2; REDD2; REDD-2; regulated in development and DNA damage response 2; RTP801L; skeletal muscle hindlimb suspension 1; Smhs1; Soleus muscle atrophied after hindlimb suspension protein 1
Gensymboler DDIT4L
Værtsarter Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DDT4L.
Oprensningsmetode Affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 115265
Målarter Human
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.