Få mere at vide
DCVC, MedChemExpress
Artikelnummer. 16497858
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16497858 10 mg
16473807 50 mg
16467676 100 mg
16430328 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16497858 Leverandør Medchem Express Leverandørnr. HY19717
MedChemExpress DCVC (S-[(1E)-1,2-dichloroethenyl]--L-cysteine) is a bioactive metabolite of trichloroethylene (TCE). DCVC inhibits pathogen-stimulated pro-inflammatory cytokines IL-1β, IL-8, and TNF-α release from tissue cultures.
DCVC (S-[(1E)-1,2-dichloroethenyl]--L-cysteine) is a bioactive metabolite of trichloroethylene (TCE). DCVC inhibits pathogen-stimulated pro-inflammatory cytokines IL-1β, IL-8, and TNF-α release from tissue cultures.
Tekniske data
Kemisk navn eller materiale DCVC
Fysisk form Solid
CAS 13419-46-0
Farve Off-White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C5H7Cl2NO2S
Synonym S-[(1E)-1,2-dichloroethenyl]--L-cysteine
Opløselighedsinformation DMSO : 2.5 mg/mL (11.57 mM; Need ultrasonic) ∣H2O : 1.034 mg/mL (4.79 mM; Need ultrasonic and warming)
SMIL N[C@H](C(O)=O)CS/C(Cl)=C\Cl
Molekylvægt (g/mol) 216.09
Formel vægt 216.09
Procent renhed 99.71%