Få mere at vide
DC_517, MedChemExpress
Artikelnummer. 16471517
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16471517 50 mg
16448027 5 mg
16415318 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16471517 Leverandør Medchem Express Leverandørnr. HY12747
MedChemExpress DC_517 is a DNA methyltransferase 1 (DNMT1) inhibitor, with an IC50 and a Kd of 1.7 μM and 0.91 μM, respectively.
DC_517 is a DNA methyltransferase 1 (DNMT1) inhibitor, with an IC50 and a Kd of 1.7 μM and 0.91 μM, respectively.
Tekniske data
Kemisk navn eller materiale DC_517
CAS 500017-70-9
Farve White
Renhedsgrad noter Research
Molekylær formel C33H35N3O2
Mængde 50 mg
Opløselighedsinformation DMSO : ≥ 50 mg/mL (98.88 mM)
SMIL OC(CNC(C)C)COC(CN1C(C=CC=C2)=C2C3=C1C=CC=C3)CN(C4=CC=CC=C45)C6=C5C=CC=C6
Molekylvægt (g/mol) 505.65
Formel vægt 505.65
Procent renhed 99.41%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death