Få mere at vide
Tocris Bioscience™ DC 260126
GREENER_CHOICE
FFA1 (GPR40) antagonist
Brand:  Tocris Bioscience™ 5357/10
Encompass
Produktkode 15766746
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
DC 260126 is a free fatty acid receptor 1 (FFA1/GPR40) antagonist. Inhibits FFA-induced increases in intracellular Ca2+ levels and suppresses palmitic acid potentiated glucose-stimulated insulin secretion in Min6 pancreatic β cells in vitro. Decreases serum insulin levels, improves insulin sensitivity and reduces the number of apoptotic pancreatic β cells in obese diabetic (db/db) rats.
Tekniske data
Tekniske data
N-(4-Butylphenyl)-4-fluorobenzenesulfonamide
346692-04-4
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner