Få mere at vide
Tocris Bioscience™ DC 260126
FFA1 (GPR40) antagonist
Brand: Tocris Bioscience™ 5357/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DC 260126 is a free fatty acid receptor 1 (FFA1/GPR40) antagonist. Inhibits FFA-induced increases in intracellular Ca2+ levels and suppresses palmitic acid potentiated glucose-stimulated insulin secretion in Min6 pancreatic β cells in vitro. Decreases serum insulin levels, improves insulin sensitivity and reduces the number of apoptotic pancreatic β cells in obese diabetic (db/db) rats.Tekniske data
| N-(4-Butylphenyl)-4-fluorobenzenesulfonamide | |
| 346692-04-4 | |
| 50 mg |