Få mere at vide
Tocris Bioscience™ DBHDA
Beskrivelse
DBHDA is a reagent for synthetic biology; converts cysteine to dehydroalanine (Dha).
Tekniske data
Tekniske data
| CAS | 99584-96-0 |
| Mængde | 50 mg |
| Molekylær formel | C6H10Br2N2O2 |
| Formel vægt | 301.96g/mol |
| Produkttype | Synthetic Biology Reagent |
For Research Use Only