Få mere at vide
Tocris Bioscience™ DBHDA
Reagent for synthetic biology; converts cysteine to dehydroalanine
Brand: Tocris Bioscience™ 6175/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DBHDA is a reagent for synthetic biology; converts cysteine to dehydroalanine (Dha).Tekniske data
| Reagent for Synthetic Biology, Converts Cysteine to Dehydroalanine | |
| C6H10Br2N2O2 | |
| Store at 4°C | |
| 50 mg |
| 99584-96-0 | |
| 301.96g/mol | |
| Buffers, Chelators and Reagents |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only