Få mere at vide
DBCO-PEG5-NHS ester, TRC
Artikelnummer. 30364764
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364764 1 mg
30355288 10 mg
30351191 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364764 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1949981MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1378531-80-6
Molekylær formel C36H43N3O11
Tekniske data
Certificeringer/compliance ISO 9001
Molekylvægt (g/mol) 693.74