missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ DBCO PEG4 Biotin
Klik for at se tilgængelige muligheder
Mængde:
10 mg
Beskrivelse
DBCO-PEG4-Biotin is a biotinylation reagent for labeling azide-containing biomolecules via Cu(I)-free Strain-Promoted Alkyne-Azide Click Chemistry (SPAAC) reaction. The hydrophilic PEG4 linker reduces or eliminates aggregation and precipitation during the labeling process by increasing the hydrophilicity of the target molecule. The PEG linker in DBCO-PEG4-Biotin also enhances the accessibility of the biotin moiety, improving the detection efficiency of the biotinylated molecule via fluorescent or HRP-labeled streptavidin or its affinity purification via streptavidin agarose.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (3aS,4S,6aR)-N-[19-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)-15,19-dioxo-3,6,9,12-tetraoxa-16-azanonadec-1-yl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide |
| CAS | 1255942-07-4 |
| Mængde | 10 mg |
| Mål | General Laboratory Research Reagent |
| Molekylær formel | C39H51N5O8S |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion