Få mere at vide
Dazomet, TRC
Artikelnummer. 30361142
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361142 1 g
30356062 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361142 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1949501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 533-74-4
Molekylær formel C5 H10 N2 S2
Molekylvægt (g/mol) 162.28
Synonym Tetrahydro-3,5-dimethyl-2H-1,3,5-thiadiazine-2-thione, 2-Thio-3,5-dimethyltetrahydro-1,3,5-thiadiazine, 3,5-Dimethyl-2-thionotetrahydro-1,3,5-thiadiazine, 3,5-Dimethyltetrahydro-1,3,5-2H-thiadiazine-2-thione, 3,5-Dimethyltetrahydro-1,3,5-thiadiazine-2-thione, 3,5-Dimethyltetrahydro-2H-1,3,5-thiadiazine-2-thione, AMA 424, Basamid, Basamid G, Basamid Granular, Basamid P, Basamid-Puder, Busan, Busan 1058, Crag 85W, Crag 974, D 35, DMTT, Dazomet, Dimethylformocarbothialdine, Fennosan B 100, Metasol D 3TA, Mylone, Mylone 85, N 521, NSC 4737, Nalco 5787, Nalcon 243, Nefusan, Nuosept S, Stauffer N 521, Tetrahydro-2H-3,5-dimethyl-1,3,5-thiadiazine-2-thione, Tetrahydro-3,5-dimethyl-1,3,5-thiadiazin-2-thione, Tetrahydro-3,5-dimethyl-2H-1,3,5-thiodiazin-2-thione, Thiazon, Thiazone, UCC 974
IUPAC navn 3,5-dimethyl-1,3,5-thiadiazinane-2-thione
SMIL CN1CSC(=S)N(C)C1
Tekniske data
Kemisk navn eller materiale Dazomet
Mængde 1 g
InChI formel InChI=1S/C5H10N2S2/c1-6-3-7(2)5(8)9-4-6/h3-4H2,1-2H3
Formel vægt 162.0285