Få mere at vide
Dapsone Hydroxylamine-d4, TRC
Artikelnummer. 30359292
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30359292 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359292 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19326210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dapsone Hydroxylamine-d4
CAS 1330185-26-6
Molekylær formel C12H8D4N2O3S
Mængde 10 mg
Synonym N-Hydroxy-4,4’-sulfonyldi(aniline)-d4
InChI formel InChI=1S/C12H12N2O3S/c13-9-1-5-11(6-2-9)18(16,17)12-7-3-10(14-15)4-8-12/h1-8,14-15H,13H2/i1D,2D,5D,6D
SMIL NC1=C([2H])C([2H])=C(S(C2=CC=C(NO)C=C2)(=O)=O)C([2H])=C1[2H]
IUPAC navn N-[4-(4-amino-2,3,5,6-tetradeuteriophenyl)sulfonylphenyl]hydroxylamine
Molekylvægt (g/mol) 268.32