Få mere at vide
Dapsone-D8 (major), TRC
Artikelnummer. 30349478
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30349478 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349478 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19325510MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dapsone-D8 (major)
CAS 557794-38-4
Molekylær formel C12 2H8 H4 N2 O2 S
Mængde 10 mg
Synonym Dapsone D8
InChI formel InChI=1S/C12H12N2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H,13-14H2/i1D,2D,3D,4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)c2c([2H])c([2H])c(N)c([2H])c2[2H]
IUPAC navn 4-(4-amino-2,3,5,6-tetradeuteriophenyl)sulfonyl-2,3,5,6-tetradeuterioaniline
Molekylvægt (g/mol) 256.35
Formel vægt 256.1122