Få mere at vide
Dapivirine-d11, TRC
Artikelnummer. 30179401
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179401 1 mg
30179112 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179401 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1855021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1329613-10-6
Molekylær formel C20H8D11N5
Molekylvægt (g/mol) 340.47
Synonym 4-[[4-[(2,4,6-Trimethylphenyl-d11)amino]-2-pyrimidinyl]amino]benzonitrile, R 147681-d11, TMC 120-d11, TMC 120R147681-d11;
IUPAC navn 4-[[4-[3,5-dideuterio-2,4,6-tris(trideuteriomethyl)anilino]pyrimidin-2-yl]amino]benzonitrile
SMIL [2H]C1=C(C([2H])([2H])[2H])C(NC2=NC(NC3=CC=C(C#N)C=C3)=NC=C2)=C(C([2H])([2H])[2H])C([2H])=C1C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale Dapivirine-d11
Mængde 1 mg
InChI formel InChI=1S/C20H19N5/c1-13-10-14(2)19(15(3)11-13)24-18-8-9-22-20(25-18)23-17-6-4-16(12-21)5-7-17/h4-11H,1-3H3,(H2,22,23,24,25)/i1D3,2D3,3D3,10D,11D