Få mere at vide
Dapagliflozin Methyl Acetate, TRC
Artikelnummer. 30354773
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354773 10 mg
30351945 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354773 Leverandør Toronto Research Chemicals Leverandørnr. TRCD18361010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dapagliflozin Methyl Acetate
CAS 1632287-34-3
Molekylær formel C23 H27 Cl O7
Mængde 10 mg
Synonym ((2R,3S,4R,5R,6S)-6-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)methyl Acetate, Dapagliflozin Impurity-S2D(Mono acetylated)
InChI formel InChI=1S/C23H27ClO7/c1-3-29-17-7-4-14(5-8-17)10-16-11-15(6-9-18(16)24)23-22(28)21(27)20(26)19(31-23)12-30-13(2)25/h4-9,11,19-23,26-28H,3,10,12H2,1-2H3/t19-,20-,21+,22-,23+/m1/s1
SMIL CCOc1ccc(Cc2cc(ccc2Cl)[C@@H]3O[C@H](COC(=O)C)[C@@H](O)[C@H](O)[C@H]3O)cc1
IUPAC navn [(2R,3S,4R,5R,6S)-6-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-3,4,5-trihydroxyoxan-2-yl]methyl acetate
Molekylvægt (g/mol) 450.909
Formel vægt 450.145