Få mere at vide
Dapagliflozin-d5, TRC
Artikelnummer. 30361687
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30361687 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361687 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1853721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dapagliflozin-d5
CAS 1204219-80-6
Renhedsgrad noter HPLC
Molekylær formel C21H20D5ClO6
Mængde 1 mg
Synonym (1S)-1,5-Anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-D-glucitol-d5, BMS 512148-D5, (2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-(ethoxy-d5)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
InChI formel InChI=1S/C21H25ClO6/c1-2-27-15-6-3-12(4-7-15)9-14-10-13(5-8-16(14)22)21-20(26)19(25)18(24)17(11-23)28-21/h3-8,10,17-21,23-26H,2,9,11H2,1H3/t17-,18-,19+,20-,21+/m1/s1/i1D3,2D2
SMIL ClC(C=CC([C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O1)=C2)=C2CC3=CC=C(OC([2H])([2H])C([2H])([2H])[2H])C=C3
IUPAC navn (2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-(ethoxy-d5)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Molekylvægt (g/mol) 413.9
Formel vægt 413.17
Procent renhed >95