Få mere at vide
Dansylamidoethyl Methanethiosulfonate, TRC
Artikelnummer. 30368782
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368782 5 mg
30353049 10 mg
30359417 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368782 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1735005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 355115-41-2
Molekylær formel C15 H20 N2 O4 S3
Molekylvægt (g/mol) 388.53
IUPAC navn 5-(dimethylamino)-N-(2-methylsulfanylsulfonylethyl)naphthalene-1-sulfonamide
SMIL CSS(=O)(=O)CCNS(=O)(=O)c1cccc2c(cccc12)N(C)C
Tekniske data
Kemisk navn eller materiale Dansylamidoethyl Methanethiosulfonate
Mængde 5 mg
InChI formel InChI=1S/C15H20N2O4S3/c1-17(2)14-8-4-7-13-12(14)6-5-9-15(13)24(20,21)16-10-11-23(18,19)22-3/h4-9,16H,10-11H2,1-3H3
Formel vægt 388.0585