missing translation for 'onlineSavingsMsg'
Learn More

Dansyl-morpholine, TRC

Product Code. 30359141
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30359141 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359141 Supplier Toronto Research Chemicals Supplier No. TRCD18030025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Dansyl-morpholine
CAS 78323-98-5
Molekylær formel C16H20N2O3S
Mængde 25 mg
Synonym DNS-morpholine, N,N-Dimethyl-5-(4-morpholinylsulfonyl)-1-naphthalenamine, 4-[[5-(Dimethylamino)-1-naphthalenyl]sulfonyl]-morpholine
InChI formel InChI=1S/C16H20N2O3S/c1-17(2)15-7-3-6-14-13(15)5-4-8-16(14)22(19,20)18-9-11-21-12-10-18/h3-8H,9-12H2,1-2H3
SMIL CN(C)c1cccc2c(cccc12)S(=O)(=O)N3CCOCC3
IUPAC navn N,N-dimethyl-5-morpholin-4-ylsulfonylnaphthalen-1-amine
Molekylvægt (g/mol) 320.41
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.