Få mere at vide
DAMPA Ethyl Ester, TRC
Artikelnummer. 30360853
Skift visning
Mængde:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360853 5 mg
30348884 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30360853 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1365155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DAMPA Ethyl Ester
CAS 43111-51-9
Molekylær formel C17H19N7O2
Mængde 5 mg
Synonym 4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoic Acid Ethyl Ester, Ethyl p-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoate, NSC 694477
InChI formel InChI=1S/C17H19N7O2/c1-3-26-16(25)10-4-6-12(7-5-10)24(2)9-11-8-20-15-13(21-11)14(18)22-17(19)23-15/h4-8H,3,9H2,1-2H3,(H4,18,19,20,22,23)
SMIL CCOC(=O)c1ccc(cc1)N(C)Cc2cnc3nc(N)nc(N)c3n2
IUPAC navn ethyl 4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoate
Molekylvægt (g/mol) 353.38
Formel vægt 353.16