Få mere at vide
Dalfampridine-d6, TRC
Artikelnummer. 30351336
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30351336 25 mg
30349173 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30351336 Leverandør Toronto Research Chemicals Leverandørnr. TRCD11350325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dalfampridine-D6
CAS 45498-20-2
Molekylær formel C5 2H6 N2
Mængde 25 mg
Synonym 4-Pyridin-2,3,5,6-d4-amine-d2 (9CI), Dalfampridine-D6, Fampridine-D6, 4-Pyridylamine-d6
InChI formel InChI=1S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4H,(H2,6,7)/i1D,2D,3D,4D/hD2
SMIL [2H]N([2H])c1c([2H])c([2H])nc([2H])c1[2H]
IUPAC navn N,N,2,3,5,6-hexadeuteriopyridin-4-amine
Molekylvægt (g/mol) 100.15
Formel vægt 100.0908