missing translation for 'onlineSavingsMsg'
Learn More

Daimuron 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316738
Change view
Click to view available options
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30316738 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316738 Supplier Dr. Ehrenstorfer Supplier No. DREA11948000AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Daimuron (Dymron)
CAS 42609-52-9
Analyt- eller komponentnavne Daimuron (Dymron)
Forsendelsestilstand +20°C
Molekylær formel C17 H20 N2 O
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
InChI formel InChI=1S/C17H20N2O/c1-13-9-11-15(12-10-13)18-16(20)19-17(2,3)14-7-5-4-6-8-14/h4-12H,1-3H3,(H2,18,19,20)
SMIL Cc1ccc(NC(=O)NC(C)(C)c2ccccc2)cc1
IUPAC navn 1-(4-methylphenyl)-3-(2-phenylpropan-2-yl)urea
Opløsningsmiddel eller Matrix Acetonitrile
Molekylvægt (g/mol) 268.35
Mængde 1 ml
Formel vægt 268.1576 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.