Få mere at vide
Daidzein-3',5',8-d3, TRC
Artikelnummer. 30240598
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240598 1 mg
30240589 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240598 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1035011MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 220930-96-1
Molekylær formel C15 2H3 H7 O4
Molekylvægt (g/mol) 257.26
Synonym 4H-1-Benzopyran-4-one-8-d, 7-hydroxy-3-(4-hydroxyphenyl-3,5-d2)-, 7-Hydroxy-3-(4-hydroxyphenyl-3,5-d2)-4H-1-benzopyran-4-one-8-d, Daidzein-3',5',8-d3, Daidzein-3',5',8-D3
IUPAC navn 8-deuterio-3-(3,5-dideuterio-4-hydroxyphenyl)-7-hydroxychromen-4-one
SMIL [2H]c1cc(cc([2H])c1O)C2=COc3c([2H])c(O)ccc3C2=O
Tekniske data
Kemisk navn eller materiale Daidzein-3',5',8-d3
Mængde 1 mg
InChI formel InChI=1S/C15H10O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-8,16-17H/i3D,4D,7D
Formel vægt 257.0767