Få mere at vide
Dacinostat, MedChemExpress
Artikelnummer. 16495768
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
Pakningsstørrelse:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16495768 10 mg -
16461927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16495768 Leverandør Medchem Express Leverandørnr. HY13606
MedChemExpress Dacinostat is a potent HDAC inhibitor, with an IC50 of 32 nM; Dacinostat also inhibits HDAC1 with an IC50 of 9 nM, and used in cancer research.
Dacinostat is a potent HDAC inhibitor, with an IC50 of 32 nM
Tekniske data
Kemisk navn eller materiale Dacinostat
Fysisk form Solid
CAS 404951-53-7
Farve Yellow
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C22H25N3O3
Til brug med (applikation) Cancer-programmed cell death
Synonym NVP-LAQ824 LAQ824
Opløselighedsinformation DMSO : ≥ 43 mg/mL (113.32 mM)
SMIL O=C(NO)/C=C/C1=CC=C(CN(CCO)CCC2=CNC3=C2C=CC=C3)C=C1
Molekylvægt (g/mol) 379.45
Formel vægt 379.45
Procent renhed 98.0%