Få mere at vide
DACA, TRC
Artikelnummer. 30355507
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355507 2.5 mg
30354720 5 mg
30355579 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30355507 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1012002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 89459-25-6
Molekylær formel C18H19N3O
Molekylvægt (g/mol) 293.36
Synonym N-[2-(Dimethylamino)ethyl]-4-acridinecarboxamide, DACA, N-[2-(Dimethylamino)ethyl]acridine-4-carboxamide, NSC 601316
IUPAC navn N-[2-(dimethylamino)ethyl]acridine-4-carboxamide
SMIL OC(=O)CC1CCC2(CC1)OOC3(OO2)C4CC5CC3CC(C5)C4
Tekniske data
Kemisk navn eller materiale DACA
Mængde 2.5 mg
InChI formel InChI=1S/C18H19N3O/c1-21(2)11-10-19-18(22)15-8-5-7-14-12-13-6-3-4-9-16(13)20-17(14)15/h3-9,12H,10-11H2,1-2H3,(H,19,22)