Få mere at vide
Dabigatran ethyl ester hydrochloride, MedChemExpress
Artikelnummer. 16494117
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16494117 50 mg
16487956 100 mg
16458228 10 mg
16450668 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16494117 Leverandør Medchem Express Leverandørnr. HY77521
MedChemExpress Dabigatran ethyl ester hydrochloride is a potent inhibitor of ribosyldihydronicotinamide dehydrogenase (NQO2) with an IC50 value of 0.8 μM and a thrombin inhibitor.
Dabigatran ethyl ester hydrochloride is a potent inhibitor of ribosyldihydronicotinamide dehydrogenase (NQO2) with an IC50 value of 0.8 μM and a thrombin inhibitor.
Tekniske data
Kemisk navn eller materiale Dabigatran ethyl ester hydrochloride
CAS 211914-50-0
Farve White
Renhedsgrad noter Research
Molekylær formel C27H30ClN7O3
Mængde 50 mg
Opløselighedsinformation DMSO : ≥ 50 mg/mL (93.28 mM) ∣H2O : 5 mg/mL (9.33 mM; Need ultrasonic)
SMIL NC(C1=CC=C(NCC2=NC3=CC(C(N(CCC(OCC)=O)C4=CC=CC=N4)=O)=CC=C3N2C)C=C1)=N.[H]Cl
Molekylvægt (g/mol) 536.03
Formel vægt 536.03
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation