Få mere at vide
Dabigatran-d3, TRC
Artikelnummer. 30359354
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30359354 0.5 mg 0.5mg
30358478 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359354 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1000920.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dabigatran N-methyl-d3
CAS 1246817-44-6
Molekylær formel C25 2H3 H22 N7 O3
Mængde 0.5 mg
Synonym Dabigatran-(N-methyl-D3)
InChI formel InChI=1S/C25H25N7O3/c1-31-20-10-7-17(25(35)32(13-11-23(33)34)21-4-2-3-12-28-21)14-19(20)30-22(31)15-29-18-8-5-16(6-9-18)24(26)27/h2-10,12,14,29H,11,13,15H2,1H3,(H3,26,27)(H,33,34)/i1D3
SMIL [2H]C([2H])([2H])n1c(CNc2ccc(cc2)C(=N)N)nc3cc(ccc13)C(=O)N(CCC(=O)O)c4ccccn4
IUPAC navn 3-[[2-[(4-carbamimidoylanilino)methyl]-1-(trideuteriomethyl)benzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoic acid
Molekylvægt (g/mol) 474.53
Formel vægt 474.2207