Få mere at vide
Tocris Bioscience™ D609
GREENER_CHOICE
Artikelnummer. 18094152
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18094152 50 mg
18045952 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18094152 Leverandør Tocris Bioscience™ Leverandørnr. 1437/50
Selective PC-PLC inhibitor
Selective competitive phosphatidyl choline-specific phospholipase C (PC-PLC) inhibitor (Ki = 6.4 μM); antiviral and antitumor agent. Also inhibits sphingomyelin synthase. Suppresses LPS- and IFNγ-induced NO production (IC50 = 20 mg/ml) and blocks oxidative glutamate toxicity in nerve cells. Antioxidant in vivo.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83373-60-8
Molekylær formel C11H18KOS2
Tekniske data
Kemisk navn eller materiale D609
Mængde 50 mg
Formel vægt 266.46
Renhed >98%
Molekylvægt (g/mol) 269.482