Få mere at vide
d4-Succinic Acid, TRC
Artikelnummer. 30427803
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30427803 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427803 Leverandør Toronto Research Chemicals Leverandørnr. TRCS688793100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Succinic acid-d4
CAS 14493-42-6
Molekylær formel C4 D4 H2 O4
Mængde 100 mg
Synonym Butanedioic acid-d4
InChI formel InChI=1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)/i1D2,2D2
SMIL [2H]C([2H])(C(=O)O)C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3-tetradeuteriobutanedioic acid
Molekylvægt (g/mol) 122.11
Formel vægt 122.0517