missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D2-40 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ D2-40 Mouse Monoclonal Antibody.
Brand: Epredia MS-1940-RQ
Additional Details : Weight : 0.02000kg
Beschreibung
D2-40 is reactive to a 40kDa O-linked sialoglycoprotein in lymphatic endothelium, fetal testis, seminomas and dysgerminomas. D2-40 is useful in demonstrating lymphatic invasion by primary tumors and in vascular lesions. Publications suggest that D2-40 positivity in lymphovascular invasion was a more prognostic predictor of breast cancer than blood vessel invasion2. It is also considered useful in differential diagnosis of epithelioid malignant mesothelioma vs adenocarcinoma1.Host Species: Mouse
Clone: D2-40
Isotype: IgG1
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: O-linked sialoglycoprotein
Molecular Weight: 40kDa
Positive Control: Breast carcinomas, mesothelioma and colon carcinoma
Cellular Localization: Membranous/Cytoplasmic
Recommended for: Immunohistochemistry
Spezifikation
Immunohistochemistry | |
Monoclonal | |
Unconjugated | |
Mouse | |
40kDa | |
Unpurified | |
RUO | |
Human | |
Unpurified |
Membranous/Cytoplasmic | |
D2-40 | |
Prediluted antibodies which are ready-to-use for Immunohistochemistry with UltraVision Quanto Detection Systems | |
O-linked sialoglycoprotein | |
Breast carcinomas, mesothelioma and colon carcinoma | |
12 mL | |
Primary | |
Store vial at 4°C. When stored at 2-8°C, this antibody is stable for 24 months. | |
IgG1 |