Få mere at vide
D-Tyrosine-d4, TRC
Artikelnummer. 30451598
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451598 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451598 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8999721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Tyrosine-d4
CAS 944386-47-4
Molekylær formel C9 D4 H7 N O3
Mængde 1 mg
Synonym (2R)-2-Amino-3-(4-hydroxyphenyl)propanoic Acid-d4, (R)-Tyrosine-d4, D-p-Tyrosine-d4, H-D-Tyr-OH-d4, D-Tyrosine-2,3,5,6-d4, D-Tyrosine-2',3',5',6'-2H4, (2R)-2-Amino-3-(4-hydroxyphenyl-2,3,5,6-d4)propanoic acid, 4-Hydroxyphenyl-d4-D-alanine, β-(p-Hydroxyphenyl)-D-alanine-d4, D-Tyrosine-D4 (4-hydroxyphenyl-2,3,5,6-D4)
InChI formel InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m1/s1/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C[C@@H](N)C(=O)O)c([2H])c([2H])c1O
IUPAC navn (2R)-2-amino-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 185.21
Formel vægt 185.099