Få mere at vide
D-Tyrosine-d2, TRC
Artikelnummer. 30452198
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30452198 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452198 Leverandør Toronto Research Chemicals Leverandørnr. TRCT899973100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Tyrosine-d2
CAS 1202064-22-9
Molekylær formel C9 D2 H9 N O3
Mængde 100 mg
Synonym D-Tyrosine-β,β-d2 (ACI), D-Tyrosine-3,3-d2, D-(β,β-2H2)Tyrosine, D-Tyrosine-D2 (propanoic-3,3-D2 acid), (2R)-2-Amino-3-(4-hydroxyphenyl)propanoic-3,3-d2 acid, 4-Hydroxyphenyl-D-alanine-3,3-d2, β-(p-Hydroxyphenyl)-D-alanine-β,β-d2, D-Tyrosine-d2, D-4-Hydroxyphenylalanine-3,3-d2
InChI formel InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m1/s1/i5D2
SMIL [2H]C([2H])([C@@H](N)C(=O)O)c1ccc(O)cc1
IUPAC navn (2R)-2-amino-3,3-dideuterio-3-(4-hydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 183.201
Formel vægt 183.086