missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
D-tryptofan methylesterhydrochlorid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | D-Tryptophan Methyl Ester Hydrochloride |
| CAS | 14907-27-8 |
| Molekylær formel | C12 H14 N2 O2 . Cl H |
| Synonym | D-Tryptophan, methyl ester, hydrochloride (1:1), D-Tryptophan, methyl ester, monohydrochloride (9CI), Tryptophan, methyl ester, monohydrochloride, D- (8CI), (R)-Tryptophan methyl ester monohydrochloride, (R)-Tryptophane methyl ester hydrochloride, Methyl D-tryptophanate hydrochloride |
| InChI formel | InChI=1S/C12H14N2O2.ClH/c1-16-12(15)10(13)6-8-7-14-11-5-3-2-4-9(8)11;/h2-5,7,10,14H,6,13H2,1H3;1H/t10-;/m1./s1 |
| SMIL | Cl.COC(=O)[C@H](N)Cc1c[nH]c2ccccc12 |
| IUPAC navn | methyl(2R)-2-amino-3-(1H-indol-3-yl)propanoat; hydrochlorid |
| Molekylvægt (g/mol) | 254.71 |
| Mængde | 5 g |
| Formel vægt | 254.0822 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?