missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[D-Trp34] Neuropeptide Y, Human, Alfa Aesar™
For cellular and molecular biology applications
Brand: Alfa Aesar J66372.M¬
Additional Details : Weight : 0.00001kg
Specifications
4329.82 | |
Human | |
Solid | |
-30° to -10°C | |
Solid |
HPLC | |
1mg | |
RUO | |
[D-Trp34] Neuropeptide Y, human |
For Research Use Only