Få mere at vide
D-Talose-1-13C, TRC
Artikelnummer. 30414248
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414248 100 mg
30426608 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30414248 Leverandør Toronto Research Chemicals Leverandørnr. TRCT005562100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70849-29-5
Molekylær formel 13CC5H12O6
Molekylvægt (g/mol) 181.16
Synonym D(+)-Talose-13C, NSC 224293-13C
IUPAC navn (2S,3S,4S,5R)-2,3,4,5,6-pentahydroxy(1^{13}C)hexanal
SMIL OC[C@@H](O)[C@H](O)[C@H](O)[C@H](O)[13CH]=O
Tekniske data
Kemisk navn eller materiale D-Talose-1-13C
Mængde 100 mg
InChI formel InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4-,5-,6+/m1/s1/i1+1
Formel vægt 181.0667