missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Sorbitol, 91.0 to 100.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Specifications
Melting Point | 98°C to 100°C |
Packaging | Plastic bottle |
Quantity | 500 g |
Beilstein | 1721899 |
Formula Weight | 182.17 |
Percent Purity | 91.0 to 100.5% |
Physical Form | Crystalline Powder |
Assay | Dry basis |
Chemical Name or Material | D-Sorbitol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'