missing translation for 'onlineSavingsMsg'
Learn More

D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7055171
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5874-57-7
C23H19Br2N3O3S
577.29
MFCD00066121
LYRWWUCOZBTOPL-UHFFFAOYNA-N
d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci
11446470
CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1
This item is not returnable. View return policy
5874-57-7
C23H19Br2N3O3S
577.29
MFCD00066121
LYRWWUCOZBTOPL-UHFFFAOYNA-N
d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci
11446470
CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1

D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 163°C to 166°C (decomposition)
Solubility Information Soluble in ethanol and methanol.
IUPAC Name methyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
Formula Weight 155.58
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material D-Serine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'