Få mere at vide
D-Serine-2,3,3-d3, TRC
Artikelnummer. 30413600
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413600 1 mg
30424881 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413600 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2709771MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1414348-52-9
Molekylær formel C3H4D3NO3
Molekylvægt (g/mol) 108.11
Synonym D-Serine-2,3,3-d3 (ACI), (2R)-2-Amino-3-hydroxypropanoic-2,3,3-d3 acid, D-Serine-2,3,3-D3
SMIL OC([2H])([2H])[C@](N)([2H])C(O)=O
Tekniske data
Kemisk navn eller materiale D-Serine-2,3,3-d3
InChI formel InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m1/s1/i1D2,2D
Mængde 1 mg
Formel vægt 108.0614