Få mere at vide
D-Ribose-L-cysteine(Mixture of Diastereomers), TRC
Artikelnummer. 30179146
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179146 1 g
30179538 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179146 Leverandør Toronto Research Chemicals Leverandørnr. TRCR4175501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 232617-15-1
Molekylær formel C8H15NO6S
Molekylvægt (g/mol) 253.27
Synonym RibCys
IUPAC navn (4R)-2-((1R,2R,3R)-1,2,3,4-tetrahydroxybutyl)thiazolidine-4-carboxylic acid
SMIL OC[C@@H](O)[C@@H](O)[C@@H](O)C1([H])SC[C@@H](C(O)=O)N1
Tekniske data
Kemisk navn eller materiale D-Ribose-L-cysteine (Mixture of Diastereomers)
InChI formel InChI=1S/C8H15NO6S/c10-1-4(11)5(12)6(13)7-9-3(2-16-7)8(14)15/h3-7,9-13H,1-2H2,(H,14,15)/t3-,4+,5+,6+,7?/m0/s1
Mængde 1 g
Formel vægt 253.06