Learn More
Thermo Scientific Chemicals D-Propargylglycine, 97%
Click to view available options
Quantity:
1 g
5 g
Description
A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 205°C (decomposition) |
Color | White |
Solubility Information | Slightly soluble in water. |
Formula Weight | 113.12 |
Percent Purity | 97% |
Quantity | 1 g |
Odor | Odorless |
Physical Form | Powder |
Chemical Name or Material | D-Propargylglycine |