Få mere at vide
D-Proline-2,5,5-d3, CDN
Artikelnummer. 30324501
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324501 0.1 g 0.1g
30322172 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324501 Leverandør CDN Leverandørnr. CDND71200.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Proline-2,5,5-d3
CAS 1994265-09-6
Molekylær formel C5 D3 H6 N O2
Mængde 0.1 g
Synonym D-Proline-2,5,5-d3, (2 R)-Pyrrolidine-2,5,5-d3-2-carboxylic acid
InChI formel InChI=1 S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6 H,1-3H2,(H,7,8)/t4-/m1/s1/i3D2,4 D
SMIL [2 H]C1([2 H])CC[C@@]([2 H])(N1)C(=O)O
IUPAC navn (2 R)-2,5,5-trideuteriopyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 118.149
Formel vægt 118.082 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity