Få mere at vide
D-Phenylalaninol-d2, TRC
Artikelnummer. 30423811
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30423811 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423811 Leverandør Toronto Research Chemicals Leverandørnr. TRCP319517100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Phenylalaninol-d2
CAS 63386-40-3
Molekylær formel C9H11D2NO
Mængde 100 mg
Synonym (betaR)-beta-Amino-benzenepropanol-d2, D-2-Amino-3-phenyl-1-propanol-d2, ((1R)-1-Hydroxymethyl-2-phenylethyl)amine-d2, (2R)-2-Amino-3-phenyl-1-propanol-d2, (R)-(+)-2-Amino-3-phenyl-1-glycinol-d2, (R)-2-Amino-1-hydroxy-3-phenylpropane-d2, (R)-2-Benzylethanolamine-d2
InChI formel InChI=1S/C9H13NO/c10-9(7-11)6-8-4-2-1-3-5-8/h1-5,9,11H,6-7,10H2/t9-/m1/s1/i7D2
SMIL [2H]C([2H])(O)[C@H](N)Cc1ccccc1
IUPAC navn (2R)-2-amino-1,1-dideuterio-3-phenylpropan-1-ol
Molekylvægt (g/mol) 153.22
Formel vægt 153.1123