Få mere at vide
D-Phenyl-d5-alanine-N-FMOC, CDN
Artikelnummer. 30326127
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326127 0.25 g -
30327283 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326127 Leverandør CDN Leverandørnr. CDND76770.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine-2,3,4,5,6-D5
CAS 2714483-74-4
Molekylær formel C24 D5 H16 N O4
Mængde 0.25 g
Synonym N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-phenyl-d5-alanine, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine-2,3,4,5,6-D5, (2 R)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]-3-(phenyl-d5)propanoic acid, D-Phenyl-d5-alanine-N-FMOC, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine-d5 (phenyl-d5), N-(9-Fluorenylmethoxycarbonyl)-D-phenyl-d5-alanine, N-Fmoc-D-phenylalanine-d5
InChI formel InChI=1 S/C24H21NO4/c26-23(27)22(14-16-8-2-1-3-9-16)25-24(28)29-15-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h1-13,21-22 H,14-15H2,(H,25,28)(H,26,27)/t22-/m1/s1/i1D,2 D,3 D,8 D,9 D
SMIL [2 H]c1c([2 H])c([2 H])c(C[C@@H](NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O)c([2 H])c1[2 H]
IUPAC navn (2 R)-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid
Molekylvægt (g/mol) 392.4587
Formel vægt 392.1784 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity